Skip to Content

BDPH9BCDCBCA

Bide Pharmatech

1-(2-Bromo-3-chlorophenyl)ethanone

Synonym(s):

1-(2-bromo-3-chlorophenyl)ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6O1Cl1Br1
CAS Number:
Molecular Weight:
233.49
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrClO/c1-5(11)6-3-2-4-7(10)8(6)9/h2-4H,1H3

Inchi Key

KCCOVPPONJFFDS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?

Explore all of our products under 1-(2-Bromo-3-chlorophenyl)ethanone


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library