Skip to Content
Merck

Skip To

BDPH9BCDE03A

Bide Pharmatech

2-Oxo-2,3-dihydrobenzo[d]oxazole-5-carboxylic acid

Synonym(s):

2-oxo-2,3-dihydro-1,3-benzoxazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C8H5O4N1
CAS Number:
Molecular Weight:
179.13
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5NO4/c10-7(11)4-1-2-6-5(3-4)9-8(12)13-6/h1-3H,(H,9,12)(H,10,11)

Inchi Key

KHVGBYCMDLCIAF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library