Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H18N2O.2ClH/c9-4-1-5-10-6-2-8(11)3-7-10;;/h8,11H,1-7,9H2;2*1H
Inchi Key
DPJKYAKTKDWSEO-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 1-(3-Aminopropyl)piperidin-4-ol dihydrochloride
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
