Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H7BrN2/c1-6-7(9)2-3-8-10-4-5-11(6)8/h2-5H,1H3
Inchi Key
UGRZKLCCPTTZKM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
White to Yellow Solid
Still not finding the right product?
Explore all of our products under 6-Bromo-5-methylimidazo[1,2-a]pyridine
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
