Skip to Content
Merck

BDPH9BCDF2F2

Bide Pharmatech

7-Hydroxyquinoline-8-carboxylic acid

Synonym(s):

7-hydroxy-8-quinolinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7O3N1
CAS Number:
Molecular Weight:
189.17
MDL number:
Product BDPH9BCDF2F2 is not currently sold in your country.


Inchi Code

1S/C10H7NO3/c12-7-4-3-6-2-1-5-11-9(6)8(7)10(13)14/h1-5,12H,(H,13,14)

Inchi Key

RQCGQULJYVNAQX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library