Skip to Content

BDPH9BCDFD7A

Bide Pharmatech

2-Methoxyacetamide

Synonym(s):

2-methoxyacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C3H7O2N1
Molecular Weight:
89.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C3H7NO2/c1-6-2-3(4)5/h2H2,1H3,(H2,4,5)

Inchi Key

MTEZLAATISORQK-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library