Skip to Content
Merck

BDPH9C01EDC8

Bide Pharmatech

(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride

Synonym(s):

(2S,4S)-4-fluoro-2-pyrrolidinecarboxamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H10O1N2Cl1F1
CAS Number:
Molecular Weight:
168.6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: solid

Inchi Code

1S/C5H9FN2O.ClH/c6-3-1-4(5(7)9)8-2-3;/h3-4,8H,1-2H2,(H2,7,9);1H/t3-,4-;/m0./s1

Inchi Key

YIEACUBSPQGCLW-MMALYQPHSA-N



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library