Skip to Content
Merck

BDPH9C021FFB

Bide Pharmatech

(R)-10-Benzyl-23-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-6,9,12,15,18-pentaoxo-3-oxa-5,8,11,14,17-pentaazatricosanoic acid

Synonym(s):

(R)-10-benzyl-23-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-6,9,12,15,18-pentahydroxy-3-oxa-5,8,11,14,17-pentaazatricosa-5,8,11,14,17-pentaenoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C021FFB is not currently sold in your country.


Inchi Code

1S/C28H36N6O10/c35-21(9-5-2-6-12-34-25(39)10-11-26(34)40)29-14-22(36)30-16-24(38)33-20(13-19-7-3-1-4-8-19)28(43)31-15-23(37)32-18-44-17-27(41)42/h1,3-4,7-8,10-11,20H,2,5-6,9,12-18H2,(H,29,35)(H,30,36)(H,31,43)(H,32,37)(H,33,38)(H,41,42)/t20-/m1/s1

Inchi Key

SODPQQOBNODMSG-HXUWFJFHSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library