Skip to Content

BDPH9C024751

Bide Pharmatech

(E)-4,4,4-Trifluoro-N-methoxy-N-methylbut-2-enamide

Synonym(s):

(2E)-4,4,4-trifluoro-N-methoxy-N-methyl-2-butenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O2N1F3
CAS Number:
Molecular Weight:
183.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8F3NO2/c1-10(12-2)5(11)3-4-6(7,8)9/h3-4H,1-2H3/b4-3+

Inchi Key

SXKYBBUJTGYOIJ-ONEGZZNKSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library