Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Skip To

PG9H9BC7AFD8

PrecisePEG

Fmoc-Ala-PAB-PNP

Synonym(s):

(9H-fluoren-9-yl)methyl hydrogen (S)-(1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxopropan-2-yl)carbonimidate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C32H27N3O8/c1-20(33-31(37)41-19-29-27-8-4-2-6-25(27)26-7-3-5-9-28(26)29)30(36)34-22-12-10-21(11-13-22)18-42-32(38)43-24-16-14-23(15-17-24)35(39)40/h2-17,20,29H,18-19H2,1H3,(H,33,37)(H,34,36)/t20-/m0/s1

Inchi Key

ODXKCZQROHNIJD-FQEVSTJZSA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



hstatements

pcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library