Skip to Content
Merck

Skip To

ONCOMLS

Oncometabolite Library of Standards

(Supplied by IROA Technologies)

Synonym(s):

Cancer Metabolite Library, Metabolite Library, Oncometabolite Library of Standards

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

IROA Technologies

suitability

suitable for LC-MS, suitable for mass spectrometry

application(s)

metabolomics
(lipidomics)

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−20°C

General description

ONCOMLS (Oncometabolite Library of Standards) is a collection of 96 high-purity (>95%) small molecule metabolite standards supplied in an economical, ready-to-use format. These compounds are specifically curated to represent key oncometabolites — a class of intracellular metabolic signaling molecules produced by both cancer cells and non-cancer cells within the tumor microenvironment that are directly implicated in tumor growth, survival, immune evasion, and epigenetic reprogramming. The ONCOMLS library provides researchers with a rigorously curated, analytically validated set of authentic metabolite standards that span the major classes of oncologically relevant small molecules — including amines, amino acids and peptides, bile acids and sterols, carbohydrates, carboxylic acids, fatty acids and acyls, nucleosides, and steroids. Each compound is provided as a pre-weighed, dried standard at 5 µg per well, offering sufficient material for multiple mass spectrometry injections. The library is formatted in a single 96-well polypropylene deep-well plate with alphanumeric assigned positions, enabling seamless integration with both manual and automated liquid handling workflows.

Application

Suitable for manual and automated work flows. Metabolite libraries are most commonly used to provide retention times and spectra for key metabolic compounds, and help optimize analytical mass spectrometry protocols with a focus on metabolite research and metabolomic studies.

Features and Benefits

1. These standards are used to provide retention times and in creation of spectra library.


2. Aids in optimizing the mass spectrometry analytical protocols, to qualify and quantify mass spectrometry sensitivity.


3. Consists of high purity compounds that are supplied in an economical, 96-well plate.


4. Pre-weighed, and supplied in dry and ready to use format.


5. Available plate map of compounds and descriptors provided in a downloadable spreadsheet below:

Preparation Note

Spin down the contents of the 96-well plate in a 96-well plate holder centrifuge at 3,000 rpm for 2–3 minutes to ensure all dried material is deposited at the bottom of each well prior to reconstitution.
Compounds in Plate 1, Rows A–D can be solubilized using ethanol; compounds in Rows E–H are water soluble. Pipette liquid up and down in the well 2–3 times to facilitate complete solubilization.
The compounds can either be used as standards and injected individually or mixed in such a way that the entire library may be examined with reasonable efficiency.

Legal Information

MLSDiscovery is a trademark of IROA Technologies LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
OAMLSGUTMLSPPMLS
description

(Supplied by IROA Technologies)

description

Supplied by IROA Technologies

description

Supplied by IROA Technologies

description

PPMLS, Polyphenol library standards, Polyphenol metabolite library

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

application(s)

metabolomics
(lipidomics)

application(s)

-

application(s)

advanced drug delivery

application(s)

advanced drug delivery
lipidomics
metabolomics
microbiology

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

relevant disease(s)

cancer

relevant disease(s)

-

relevant disease(s)

-

relevant disease(s)

-

suitability

suitable for mass spectrometry

suitability

-

suitability

-

suitability

suitable for mass spectrometry, suitable for (metabolite standards)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library