Skip to Content
Merck

SML4002

SGC-CAF382-1

≥98% (HPLC)

Synonym(s):

CAF 382, CAF 382 (B1), CAF-382, CAF-382 (B1), CAF382, CAF382 (B1), N-(5-(((5-Isopropyloxazol-2-yl)methyl)thio)thiazol-2-yl)piperidine-4-carboxamide 2,2,2-trifluoroacetate

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=NC=C(S1)SCC2=NC=C(O2)C(C)C)C3CCNCC3.[xTFA]

Biochem/physiol Actions

Potent cyclin-dependent kinase-like 5 inhibitor with additional potency against CDK9/16/17/18 and good kinome-wide selectivity.
SGC-CAF382-1 is a potent cyclin-dependent kinase-like 5 inhibitor (CDKL5 IC50 = 6.7 nM by binding assay & 10 nM by cellular NanoBRET) with additional potency against CDK9/16/17/18 (IC50 = 20/62/89/100 nM by cell-free enzymatic assay, 280/390/240/260 nM by cellular NanoBRET) and good kinome-wide selectivity. SGC-CAF382-1 downregulates EB2 Ser222 phosphorylation in DIV14-16 rat primary neurons (IC50 <5 nM) and in hippocampal slices from P20–30 rats ex vivo (IC50 <45 nM).

Disclaimer

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Castano et al.
eLife, 12 (2023-07-25)
Pathological loss-of-function mutations in cyclin-dependent kinase-like 5 (CDKL5) cause CDKL5 deficiency disorder (CDD), a rare and severe neurodevelopmental disorder associated with severe and medically refractory early-life epilepsy, motor, cognitive, visual, and autonomic disturbances in the absence of any structural brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service