Merck
Z744853
Squib Separatory Funnel with Stopcock
new
capacity 2000 mL, borosilicate glass
Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41121809
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PTFE stopcock
borosilicate glass
Joint
Joints stopcock (6)
joint stopper29/42
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 1426T-2000
capacity
2000 mL
General description
The Squibb Style Separatory Funnel with stopper features a smooth tooled tip, large white block letters, and large marking squares.
Features and Benefits
  • Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
  • Made from high-quality borosilicate tubing with a low coefficient of expansion
  • Stopper No.: 29/42
  • Stopcock Plug Size: 6
Choose from one of the most recent versions:
Certificates of Analysis (COA)