Skip to Content
Merck

33053

2,2-Dimethoxypropane

Synonym(s):

DMP, Acetone dimethyl acetal

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Linear Formula:
(CH3)2C(OCH3)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
635678
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

derivatization grade ((GC derivatization))

vapor density

3.59 (vs air)

vapor pressure

60 mmHg ( 15.8 °C)

description

Alkylation reagent

expl. lim.

31 %, 58 °F
6 %, 27 °F

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

bottle of 25 g

refractive index

n20/D 1.378 (lit.)

bp

83 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)OC

InChI

1S/C5H12O2/c1-5(2,6-3)7-4/h1-4H3

InChI key

HEWZVZIVELJPQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for fatty acid methyl ester, isopropylidene glycerol.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Goldstein
Scanning Electron Microscopy and X-Ray Microanalysis: A Text for Biologists, Materials Scientists, and Geologists, 587-587 (2012)
H Shimasaki et al.
Journal of lipid research, 18(4), 540-543 (1977-07-01)
A method is described for the transesterification of lipids of mammalian tissues for fatty acid analysis by gas-liquid chromatography (GLC) that eliminates the extraction step of conventional procedures. The method involves the direct reaction of anhydrous HCl-methanol with the lipids
M.E.Mason and G.R. Waller
Analytical Chemistry, 36, 583-583 (1964)
[Quantitative double-labeled autoradiography by chemical washing method using 2,2-dimethoxypropane].
H Sumiya et al.
Kaku igaku. The Japanese journal of nuclear medicine, 24(3), 315-318 (1987-03-01)
Akito Takeuchi et al.
Biomedical chromatography : BMC, 24(5), 465-471 (2009-08-19)
A method for routinely determination of dimethyl sulfoxide (DMSO) and dimethyl sulfone (DMSO(2)) in human urine was developed using gas chromatography-mass spectrometry. The urine sample was treated with 2,2-dimethoxypropane (DMP) and hydrochloric acid for efficient removal of water, which causes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service