Merck
  • Effect of chronic antipsychotic treatment on striatal phosphodiesterase 10A levels: a [¹¹C]MP-10 PET rodent imaging study with ex vivo confirmation.
Effect of chronic antipsychotic treatment on striatal phosphodiesterase 10A levels: a [¹¹C]MP-10 PET rodent imaging study with ex vivo confirmation.
Translational psychiatry (2014-04-03)
S Natesan, S Ashworth, J Nielsen, S-P Tang, C Salinas, S Kealey, J B Lauridsen, T B Stensbøl, R N Gunn, E A Rabiner, S Kapur
ABSTRACT
A number of phosphodiesterase 10A (PDE10) inhibitors are about to undergo clinical evaluation for their efficacy in treating schizophrenia. As phosphodiesterases are in the same signalling pathway as dopamine D2 receptors, it is possible that prior antipsychotic treatment could influence these enzyme systems in patients. Chronic, in contrast to acute, antipsychotic treatment has been reported to increase brain PDE10A levels in rodents. The aim of this study was to confirm these findings in a manner that can be translated to human imaging studies to understand its consequences. Positron emission tomography (PET) scanning was used to evaluate PDE10A enzyme availability, after chronic haloperidol administration, using a specific PDE10A ligand ([(11)C]MP-10). The binding of [(11)C]MP-10 in the striatum and the cerebellum was measured in rodents and a simplified reference tissue model (SRTM) with cerebellum as the reference region was used to determine the binding potential (BPND). In rats treated chronically with haloperidol (2 mg kg(-1) per day), there was no significant difference in PDE10A levels compared with the vehicle-treated group (BPND±s.d.: 3.57 ± 0.64 versus 2.86 ± 0.71). Following PET scans, ex vivo analysis of striatal brain tissue for PDE10A mRNA (Pde10a) and PDE10A enzyme activity showed no significant difference. Similarly, the PDE10A protein content determined by western blot analysis was similar between the two groups, contrary to an earlier finding. The results of the study indicate that prior exposure to antipsychotic medication in rodents does not alter PDE10A levels.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SAFC
BIS-TRIS
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Isoflurane, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard