Merck
  • GW4064, a farnesoid X receptor agonist, upregulates adipokine expression in preadipocytes and HepG2 cells.
GW4064, a farnesoid X receptor agonist, upregulates adipokine expression in preadipocytes and HepG2 cells.
World journal of gastroenterology (2014-11-18)
Xiao-Min Xin, Mu-Xiao Zhong, Gong-Li Yang, Yao Peng, Ya-Li Zhang, Wei Zhu
ABSTRACT
To investigate the effect of GW4064 on the expression of adipokines and their receptors during differentiation of 3T3-L1 preadipocytes and in HepG2 cells. The mRNA expression of farnesoid X receptor (FXR), peroxisome proliferator-activated receptor-gamma 2 (PPAR-γ2), adiponectin, leptin, resistin, adiponectin receptor 1 (AdipoR1), adiponectin receptor 2 (AdipoR2), and the long isoform of leptin receptor (OB-Rb) and protein levels of adiponectin, leptin, and resistin were determined using fluorescent real-time PCR and enzyme linked immunosorbent assay, respectively, on days 0, 2, 4, 6, and 8 during the differentiation of 3T3-L1 preadipocytes exposed to GW4064. Moreover, mRNA expression of AdipoR2 and OB-Rb was also examined using fluorescent real-time PCR at 0, 12, 24, and 48 h in HepG2 cells treated with GW4064. The mRNA expression of FXR, PPAR-γ2, adiponectin, leptin, resistin, AdipoR1, AdipoR2, and OB-Rb and protein levels of adiponectin, leptin, and resistin increased along with differentiation of 3T3-L1 preadipocytes (P < 0.05 for all). The mRNA expression of FXR, PPAR-γ2, adiponectin, leptin, and AdipoR2 in 3T3-L1 preadipocytes, and AdipoR2 and OB-Rb in HepG2 cells was significantly increased after treatment with GW4064, when compared with the control group (P < 0.05 for all). A similar trend was observed for protein levels of adipokines (including adiponectin, leptin and resistin). However, the expression of resistin, AdipoR1, and OB-Rb in 3T3-L1 cells did not change after treatment with GW4064. The FXR agonist through regulating, at least partially, the expression of adipokines and their receptors could offer an innovative way for counteracting the progress of metabolic diseases such as nonalcoholic fatty liver disease.
Product Description
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard