Merck
  • Cathelicidin peptide LL-37 increases UVB-triggered inflammasome activation: possible implications for rosacea.
Cathelicidin peptide LL-37 increases UVB-triggered inflammasome activation: possible implications for rosacea.
Journal of dermatological science (2014-10-13)
Suzanna Salzer, Sonja Kresse, Yoji Hirai, Sarah Koglin, Markus Reinholz, Thomas Ruzicka, Jürgen Schauber
ABSTRACT
In patients with rosacea, environmental stressors, especially UVB radiation, trigger disease flares that are characterized by inflammation and vascular hyperactivity. An altered innate immune detection and response system, modulated to a large extent by the aberrant production and processing of human cathelicidin LL-37, is thought to play a central role in disease pathogenesis. To investigate whether the proinflammatory and proangiogenic effects of UV radiation are enhanced in the presence of cathelicidin LL-37. Human skin ex vivo and epidermal keratinocytes in vitro were exposed to UVB irradiation. The proinflammatory effects of UVB exposure in the presence and absence of LL-37 were characterized using immunoblot, transfection, qPCR, and a cell-based second messenger assay. ELISA was used to assess cytokine release and the angiogenic potential of endothelial cells was evaluated using an in vitro angiogenesis assay. UVB irradiation triggered the inflammasome-mediated processing and release of IL-1β. LL-37 augmented this UV-induced IL-1β secretion by acting on the P2X7 receptor on keratinocytes. P2X7 receptor activation by UVB and LL-37 resulted in an increase in intracellular calcium concentrations, which enhances inflammasome activation and subsequent IL-1β release. Furthermore, IL-1β and LL-37 worked synergistically to increase the angiogenic potential of endothelial cells. Cathelicidin LL-37 modulates the proinflammatory and proangiogenic effects of UV radiation and thereby contributes to enhanced sensitivity to sun exposure in rosacea.
Sigma-Aldrich
Calcium Ionophore A23187 mixed calcium magnesium salt, Approximate 1:15 molar ratio Ca:Mg.
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
KN-62, ≥95%, powder
SAFC
BIS-TRIS
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%