Merck
  • Metabolomics of the aqueous humor in the rat glaucoma model induced by a series of intracamerular sodium hyaluronate injection.
Metabolomics of the aqueous humor in the rat glaucoma model induced by a series of intracamerular sodium hyaluronate injection.
Experimental eye research (2014-12-06)
A Mayordomo-Febrer, M López-Murcia, J M Morales-Tatay, D Monleón-Salvado, M D Pinazo-Durán
ABSTRACT
Glaucoma models are helpful to study disease characteristics and to design new therapeutic options. Metabolomic profiling approach have been used to elucidating the molecular characteristics of the aqueous humor. Juvenile male Wistar rats experimental (n = 15) and controls (n = 6) were used for these studies. Experimental rats received weekly intracamerular injection of 25 µl of sodium hyaluronate in the left eye and sterile saline solution in the right eye, consecutively for ten weeks. Rats were subjected to anterior/posterior eye segment examinations, intraocular pressure (IOP), and flash electroretinograms (ERG). The aqueous humor was collected at endpoints and analyzed by Nuclear Magnetic Resonance. Elevated IOP and significant reduction of a, b waves and amplitude of oscillatory potential was observed in the left eyes compared to control eyes. The aqueous humor metabolomic profile from control and the experimental eyes were compared. Concentrations of metabolites (amino acids, lipids and carbohydrates) significantly changed after the sodium hyaluronate injections series, compared to the sham-operated eyes. Metabolic changes in the hypertensive eyes correlated with the impaired retinal function. Observed metabolomic changes in aqueous humor in hypertensive state may play a significant role in glaucoma pathogenesis.
Product Description
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Tropicamide, solid
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)