Merck
  • Injectable microcarriers as human mesenchymal stem cell support and their application for cartilage and degenerated intervertebral disc repair.
Injectable microcarriers as human mesenchymal stem cell support and their application for cartilage and degenerated intervertebral disc repair.
European cells & materials (2015-01-13)
A Bertolo, S Häfner, A R Taddei, M Baur, T Pötzel, F Steffen, J Stoyanov
ABSTRACT
Degeneration of the intervertebral disc (IVD) is a progressive and chronic process, and the high incidence of discogenic disorders calls for new therapeutic approaches, such as cell-based therapies using three dimensional cultures and mesenchymal stem cells (MSC), which can differentiate to chondrogenic- and IVD-lineages. Here, we investigated the growth and differentiation of human MSC culture on biodegradable collagen scaffolds in order to obtain an injectable suspension. Commercially available wound dressings were downsized to dimensions between 100 and 1500 μm and seeded with freshly isolated or early passages MSC. Proliferation rate and chondrogenic differentiation potential was tested at oxygenation levels of 2%, 5%, 10% and 21% in static and dynamic cultures. Evaluation methods included cell viability test, disc marker genes expression (aggrecan, collagen type I and type II), histological detection of proteoglycans and immunohistochemical analysis. On microcarriers, freshly isolated MSC had lower proliferation rate and chondrogenic differentiation potential compared with early passages MSC. Proliferation of MSC was significantly increased 1.7-fold at 5% oxygen level and in combination with dynamic culture was further increased to 2.3-fold, with respect to normoxia. Chondrogenesis was positively affected by 2% and 5% hypoxia, as shown by increased transcription levels and protein expression of collagen type II and proteoglycan accumulation in static cultures, while it was inhibited in dynamic cultures. Collagen type I and aggrecan expression were not affected by hypoxia. In conclusion, collagen based microcarriers are a suitable support for in vitro MSC growth and chondrogenesis especially when cultured at 5% oxygen level.
Sigma-Aldrich
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
SAFC
L-Proline
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis