Merck
  • The anti-inflammatory secoiridoid glycosides from gentianae scabrae radix: the root and rhizome of Gentiana scabra.
The anti-inflammatory secoiridoid glycosides from gentianae scabrae radix: the root and rhizome of Gentiana scabra.
Journal of natural medicines (2015-03-10)
Yu-Min He, Shu Zhu, Yue-Wei Ge, Kohei Kazuma, Kun Zou, Shao-Qing Cai, Katsuko Komatsu
ABSTRACT
Gentianae Scabrae Radix is a well-known traditional medicine that is used for the treatment of hepatitis, cholecystitis and inflammatory diseases. It consists mainly of secoiridoid glycosides, with representatives of gentiopicroside, sweroside and swertiamarin. In the present study, a chemical investigation of the CHCl3 extract of Gentianae Scabrae Radix derived from Gentiana scabra Bunge yielded seven new (1-7) and ten known (8-17) secoiridoid glycosides. Their structures were elucidated by extensive spectroscopic analyses and comparison with literature data. All 17 compounds were evaluated for their inhibitory effects against NO, IL-6 and TNF-α productions induced by lipopolysaccharide (LPS) in RAW264 cells. Among them, 8-epi-kingiside derivatives 1-3; kingiside derivatives 4, 5 and 10; and a sweroside derivative 6 showed inhibition activity against IL-6 production with IC50 values of 51.70-61.10 μM, whereas sweroside derivatives 12 and 15-17 and a swertiamarin derivative 13 showed inhibition effects on both NO and IL-6 productions with IC50 values of 64.74-94.95 and 48.91-75.45 μM, respectively. All the compounds exhibited weak inhibitory activity (IC50 > 100 μM) in a TNF-α bioassay. Finally, a primary structure-activity relationship of these secoiridoid glycosides is discussed.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Cysteine methyl ester hydrochloride, 98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture