Merck
  • Digestibility of new dietary fibre materials, resistant glucan and hydrogenated resistant glucan in rats and humans, and the physical effects in rats.
Digestibility of new dietary fibre materials, resistant glucan and hydrogenated resistant glucan in rats and humans, and the physical effects in rats.
The British journal of nutrition (2015-10-17)
Tsuneyuki Oku, Kenichi Tanabe, Shigeki Morita, Norihisa Hamaguchi, Fumio Shimura, Sadako Nakamura
ABSTRACT
Resistant glucan (RG) and hydrogenated resistant glucan (HRG) are newly developed non-digestible carbohydrate materials that decrease lifestyle-related diseases. The bioavailability of RG and HRG was investigated by in vitro experiments using human and rat small intestinal enzymes and by in vivo experiments using rats in the present study. Oligosaccharides, which are minor components of RG and HRG, were hydrolysed slightly by small intestinal enzymes of humans and rats, and the hydrolysing activity was slightly higher in rats than in humans. The amount of glucose released from HRG was greater than that from RG. However, the high-molecular-weight carbohydrates of the main components were hardly hydrolysed. Furthermore, neither RG nor HRG inhibited disaccharidase activity. When rats were raised on a diet containing 5 % of RG, HRG, resistant maltodextrin or fructo-oligosaccharide (FOS) for 4 weeks, all rats developed loose stools and did not recover during the experiment, except for the FOS group. Body weight gain was normal in all groups and was not significantly different compared with the control group. Caecal tissue and content weights were significantly increased by feeding RG or HRG, although other organ and tissue weights were not significantly different among the groups. In conclusion, RG and HRG consist of small amounts of glucose and digestible and non-digestible oligosaccharides, and large amounts of glucose polymers, which were hardly hydrolysed by α-amylase and small intestinal enzymes. RG and HRG, which were developed newly as dietary fibre materials, had no harmful effects on the growth and development of rats.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline sodium salt
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications