Merck
Effect of resveratrol on experimental non-alcoholic steatohepatitis.
Pharmacological research (2015-03-31)
Sara Heebøll, Karen Louise Thomsen, Andrew Clouston, Elias Immanuel Sundelin, Yulia Radko, Lars Porskjær Christensen, Mehdi Ramezani-Moghadam, Martin Kreutzfeldt, Steen Bønløkke Pedersen, Niels Jessen, Lionel Hebbard, Jacob George, Henning Grønbæk
ABSTRACT
Non-alcoholic fatty liver disease and non-alcoholic steatohepatitis (NASH) are increasing clinical problems for which effective treatments are required. The polyphenol resveratrol prevents the development of fatty liver disease in a number of experimental studies. We hypothesized that it could revert steatohepatitis, including hepatic inflammation and fibrosis, in an experimental NASH model. To induce hepatic steatohepatitis, a 65% fat, 2% cholesterol and 0.5% cholate (HFC) diet was fed to rats for 1 or 16 weeks, prior to treatment. Subsequently, the diet was supplemented with resveratrol (approx. 100mg/rat/day) to three intervention groups; week 2-4, 2-7 or 17-22. Treated animals were sacrificed at the end of each intervention period with appropriate control and HFC diet controls. Blood and liver were harvested for analysis. When commenced early, resveratrol treatment partially mitigated transaminase elevations, hepatic enlargement and TNFα induced protein-3 protein expression, but generally resveratrol treatment had no effect on elevated hepatic triglyceride levels, histological steatohepatitis or fibrosis. We observed a slight reduction in Collagen1α1 mRNA expression and no reduction in the mRNA expression of other markers of fibrosis, inflammation or steatosis (TGFβ, TNFα, α2-MG, or SREBP-1c). Resveratrol metabolites were detected in serum, including trans-resveratrol-3-O-sulphate/trans-resveratrol-4'-O-sulphate (mean concentration 7.9 μg/ml). Contrary to the findings in experimental steatosis, resveratrol treatment had no consistent therapeutic effect in alleviating manifest experimental steatohepatitis.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis