Cambiar Vistas
| Talla/SKU | Disponibilidad | Precio |
|---|
Acerca de este artículo
UNSPSC Code:
41105300
eCl@ss:
32160702
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarlepackaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6204-26, Cytiva TE22
General description
TE 22 Mini Tank Transfer Unit
The built-in alumina-covered heat exchanger dissipates the heat generated by tank electrotransfer. The temperature rises no more than 5°C during a typical transfer with a magnetic stirrer circulating the buffer and tap water running through the heat exchanger.
- Perform protein and nucleic acid transfers in less than 1 hour.
- Transfer four mini-gels up to 9 x 10 cm in about 1 L of buffer.
- Control transfer temperature through built-in alumina heat exchanger.
- Set up transfers quickly with the hinged, color-coded cassettes.
- To assure a uniform electrical field, Mini Tank Transfer Unit has 80 cm of wire in seven parallel strands on each electrode panel.
The built-in alumina-covered heat exchanger dissipates the heat generated by tank electrotransfer. The temperature rises no more than 5°C during a typical transfer with a magnetic stirrer circulating the buffer and tap water running through the heat exchanger.
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?