Merck
Time course of isolated rat fundus response to muscarinic agonists: a measure of intrinsic efficacy.
Physiological research (1999-08-24)
S M Jankovic, D Kouvelas, V Mirtsou-Fidani
RESUMEN
The establishment of a dose-response relationship and its quantification is the usual procedure for analysing drug action on an isolated organ. However, the time course of the effect seems to be an inherent characteristic of the agonist which produces it. In our study, we have analyzed the time-response curves of four cholinergic agonists (acetylcholine, methacholine, carbachol and bethanechol) which produce tonic contractions of the isolated rat gastric fundus. The order of affinity of agonists to muscarinic receptors on the rat fundus were carbachol > bethanechol > methacholine > acetylcholine (K(A) values: 46 +/- 12, 84 +/- 21, 380 +/- 110 and 730 +/- 120 nM, respectively). The effective concentrations which produced 60% of the maximal response (EC60) were used for establishing the time-response curves. The time-response curves were also recorded after partial alkylation of muscarinic receptors with phenoxybenzamine, after exposure of the isolated rat fundus to physostigmine and after addition of supramaximal concentrations of the agonists. The experimental time-response curve for acetylcholine was on the extreme left, followed by curves for methacholine, bethanechol and carbachol, respectively. Phenoxybenzamine and supramaximal doses of the agonists did not change the order of response development in time, but supramaximal doses shifted all curves to the left and phenoxybenzamine shifted all time-response curves to the right. Only physostigmine shifted the time-response curve for methacholine to the right. The results of our study suggest that the response rate of the isolated rat gastric fundus to cholinergic agonists depends on the intrinsic activity of these agents, but not on their affinity for muscarinic receptors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Carbamyl-β-methylcholine chloride, ≥99% (TLC), crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad