Merck
Responses of brain and behavior to changing day-length in the diurnal grass rat (Arvicanthis niloticus).
Neuroscience (2013-01-15)
G Leach, C Ramanathan, J Langel, L Yan
RESUMEN
Seasonal affective disorder (SAD) is a major depressive disorder that recurs in the fall and winter when day-length gets short. It is well accepted that day-length is encoded by the principal circadian clock located in the suprachiasmatic nucleus (SCN), but very little is known about day-length encoding in diurnal mammals. The present study utilized the grass rat, Arvicanthis niloticus, to investigate how the circadian system responds to photoperiodic changes in a diurnal mammal that shows day-length-dependent mood changes. The animals were initially housed in equatorial day-length (12h, EP) followed by either long (16h, LP) or short (8h, SP) photoperiods. The LP animals showed an expansion of the peak phase of the PER1 and PER2 rhythm in the SCN as well as an extended behavioral active phase. In contrast, the SP animals did not show any compression of their active phase nor a change in the peak duration of PER1 or PER2 expression, compared to those in EP. The results suggest that the circadian system in the diurnal grass rats is less responsive when day-length gets short compared to when it gets longer. The depression-like behaviors were assessed using sweet solution preference (SSP) and forced swimming test (FST). Animals in the SP group showed decreased SSP and increased immobility time in FST as compared to the EP group, suggesting a depressive phenotype. The present study serves as the first step toward exploring the role that the circadian system plays in SAD using a diurnal rodent model.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-mPER1 (residues 6-21) Antibody, serum, from rabbit
Sigma-Aldrich
Anticuerpo anti-PER2, serum, Chemicon®