Merck
Polycyclic aromatic triptycenes: oxygen substitution cyclization strategies.
Journal of the American Chemical Society (2012-04-19)
Brett VanVeller, Derek J Schipper, Timothy M Swager
RESUMEN
The cyclization and planarization of polycyclic aromatic hydrocarbons with concomitant oxygen substitution was achieved through acid catalyzed transetherification and oxygen-radical reactions. The triptycene scaffold enforces proximity of the alcohol and arene reacting partners and confers significant rigidity to the resulting π-system, expanding the tool set of iptycenes for materials applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-Thienylboronic acid, ≥95.0%