Merck
  • Paradoxical resistance to high-fat diet-induced obesity and altered macrophage polarization in mineralocorticoid receptor-overexpressing mice.
Paradoxical resistance to high-fat diet-induced obesity and altered macrophage polarization in mineralocorticoid receptor-overexpressing mice.
American journal of physiology. Endocrinology and metabolism (2013-11-14)
Emmanuelle Kuhn, Christine Bourgeois, Vixra Keo, Say Viengchareun, Adeline Muscat, Geri Meduri, Damien Le Menuet, Bruno Fève, Marc Lombès
RESUMEN
The mineralocorticoid receptor (MR) exerts proadipogenic and antithermogenic effects in vitro, yet its in vivo metabolic impact remains elusive. Wild type (WT) and transgenic (Tg) mice overexpressing human MR were subjected to standard chow (SC) or high-fat diet (HFD) for 16 wk. Tg mice had a lower body weight gain than WT animals and exhibited a relative resistance to HFD-induced obesity. This was associated with a decrease in fat mass, an increased population of smaller adipocytes, and an improved glucose tolerance compared with WT animals. Quantitative RT-PCR studies revealed decreased expression of PPARγ2, a master adipogenic gene, and of glucocorticoid receptor and 11β-hydroxysteroid dehydrogenase type 1, consistent with an impaired local glucocorticoid signaling in adipose tissues (AT). This paradoxical resistance to HFD-induced obesity was not related to an adipogenesis defect since differentiation capacity of Tg preadipocytes isolated from stroma-vascular fractions was unaltered, suggesting that other nonadipocyte factors might compromise AT development. Although AT macrophage infiltration was not different between genotypes, Tg mice exhibited a distinct macrophage polarization, as revealed by FACS analysis and CD11c/CD206 expression studies. We further demonstrated that Tg macrophage-conditioned medium partially impaired preadipocyte differentiation. Therefore, we propose that modification of M1/M2 polarization of hMR-overexpressing macrophages could account in part for the metabolic phenotype of Tg mice. Collectively, our results provide evidence that MR exerts a pivotal immunometabolic role by controlling adipocyte differentiation processes directly but also indirectly through macrophage polarization regulation. Our findings should be taken into account for the pharmacological treatment of metabolic disorders.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)