- The structure and function of acid proteases. Specific inactivation of an acid protease from Rhizopus chinensis by diazoacetyl-DL-norleucine methyl ester.
The structure and function of acid proteases. Specific inactivation of an acid protease from Rhizopus chinensis by diazoacetyl-DL-norleucine methyl ester.
Journal of biochemistry (1973-01-01)
F Mizobe, K Takahashi, T Ando
PMID4570372
MATERIALES