Merck
A method for genetic modification of human embryonic stem cells using electroporation.
Nature protocols (2007-04-21)
Magdaline Costa, Mirella Dottori, Koula Sourris, Pegah Jamshidi, Tanya Hatzistavrou, Richard Davis, Lisa Azzola, Steven Jackson, Sue Mei Lim, Martin Pera, Andrew G Elefanty, Edouard G Stanley
RESUMEN
The ability to genetically modify human embryonic stem cells (HESCs) will be critical for their widespread use as a tool for understanding fundamental aspects of human biology and pathology and for their development as a platform for pharmaceutical discovery. Here, we describe a method for the genetic modification of HESCs using electroporation, the preferred method for introduction of DNA into cells in which the desired outcome is gene targeting. This report provides methods for cell amplification, electroporation, colony selection and screening. The protocol we describe has been tested on four different HESC lines, and takes approximately 4 weeks from electroporation to PCR screening of G418-resistant clones.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Proteinasa K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture