Merck
  • Cloning, sequencing, and characterization of lipase genes from a polyhydroxyalkanoate (PHA)-synthesizing Pseudomonas resinovorans.
Cloning, sequencing, and characterization of lipase genes from a polyhydroxyalkanoate (PHA)-synthesizing Pseudomonas resinovorans.
Applied microbiology and biotechnology (2012-05-31)
Jeung Hee Lee, Richard D Ashby, David S Needleman, Ki-Teak Lee, Daniel K Y Solaiman
RESUMEN
Lipase (lip) and lipase-specific foldase (lif) genes of a biodegradable polyhydroxyalkanoate (PHA)-synthesizing Pseudomonas resinovorans NRRL B-2649 were cloned using primers based on consensus sequences, followed by polymerase chain reaction-based genome walking. Sequence analyses showed a putative Lip gene product (314 amino acids, a.a.) with its catalytic active site (Ser(111), Asp(258), and His(280)) identified. The foldase lif gene that is located 55 bp downstream of lip codes for a putative Lif (345 a.a.). To verify the biological function of the cloned lip gene for lipase expression in P. resinovorans, we constructed a lip knock-out mutant (lip::Tn5<KAN-2>) by transposon insertion. Complementation of the lip knock-out P. resinovorans mutant with a lipase expression plasmid (pBS29-P2-lip) was performed, and its effect on lipase expression was investigated. The wild-type P. resinovorans and the lip::Tn5<KAN-2>[pBS29-P2-lip] recombinant (but not the lip::Tn5<KAN-2> mutant) showed fluorescence on rhodamine B plates indicative of lipase activity. The wild type exhibited extracellular lipase activity when grown on medium containing triacylglycerol substrates (tallow, olive oil, and tributyrin) as sole carbon sources, but the lip::Tn5<KAN-2> mutant did not show such activity. Lipase activity of various strains was also confirmed by TLC analysis of the composition of acylglycerols and free fatty acid in the extracts of the spent culture medium. We further found that tributyrin was more effective than olive oil in inducing lipase expression in P. resinovorans.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Kits para ADN genómico bacteriano GenElute, sufficient for 10 purifications