Merck
Automated image-based phenotypic analysis in zebrafish embryos.
Developmental dynamics : an official publication of the American Association of Anatomists (2009-02-25)
Andreas Vogt, Andrzej Cholewinski, Xiaoqiang Shen, Scott G Nelson, John S Lazo, Michael Tsang, Neil A Hukriede
RESUMEN
Presently, the zebrafish is the only vertebrate model compatible with contemporary paradigms of drug discovery. Zebrafish embryos are amenable to automation necessary for high-throughput chemical screens, and optical transparency makes them potentially suited for image-based screening. However, the lack of tools for automated analysis of complex images presents an obstacle to using the zebrafish as a high-throughput screening model. We have developed an automated system for imaging and analyzing zebrafish embryos in multi-well plates regardless of embryo orientation and without user intervention. Images of fluorescent embryos were acquired on a high-content reader and analyzed using an artificial intelligence-based image analysis method termed Cognition Network Technology (CNT). CNT reliably detected transgenic fluorescent embryos (Tg(fli1:EGFP)(y1)) arrayed in 96-well plates and quantified intersegmental blood vessel development in embryos treated with small molecule inhibitors of anigiogenesis. The results demonstrate it is feasible to adapt image-based high-content screening methodology to measure complex whole organism phenotypes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
SU 4312, ≥98% (HPLC)