Merck
New glucose-6-phosphate dehydrogenase mutations associated with chronic anemia.
Blood (1995-03-01)
P J Mason, M F Sonati, D MacDonald, C Lanza, D Busutil, M Town, C M Corcoran, J S Kaeda, D J Stevens, S al-Ismail
RESUMEN
We have identified the glucose-6-phosphate dehydrogenase mutations responsible for enzyme deficiency in nine individuals with chronic nonspherocytic hemolytic anemia. We found the variants Tokyo, Iowa, Shinshu, and Guadalajara in British subjects and Kobe in an Italian. In addition we have determined the variant Corum has the mutation 820 G-->A and have found in British subjects the mis-sense mutations 224 T-->C, 488 G-->A and 833 C-->T which have not been described before. Some, but not all, of the mutations involve amino acids located near putative substrate binding sites.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret