Merck
In vitro antifungal evaluation and studies on the mode of action of xanthoxyline derivatives.
Arzneimittel-Forschung (2000-01-15)
T R Pinheiro, R A Yunes, S N López, C B Santecchia, S A Zacchino, V Cechinel Filho
RESUMEN
This study describes the fungistatic effect of xanthoxyline (CAS 90-24-4) and its derivatives against a panel of yeasts, filamentous fungi and dermatophytes, by using the agar dilution method. Results indicated that simple structural modifications led to more potent derivatives, especially in relation with dermatophytes. The most active compound tested (10), which is a benzenesulphonyl derivative, was 12-fold more potent than xanthoxyline itself against Trichophyton rubrum. The evaluation of the mode of action with the whole cell Neurospora crassa assay, suggested that some selected compounds may be acting by the inhibition of fungal cell-wall polymers synthesis or assembly.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2′-Hydroxy-4′,6′-dimethoxyacetophenone, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad