Merck
  • Evaluation of liver and peripheral blood micronucleus assays with 9 chemicals using young rats. A study by the Collaborative Study Group for the Micronucleus Test (CSGMT)/Japanese Environmental Mutagen Society (JEMS)-Mammalian Mutagenicity Study Group (MMS).
Evaluation of liver and peripheral blood micronucleus assays with 9 chemicals using young rats. A study by the Collaborative Study Group for the Micronucleus Test (CSGMT)/Japanese Environmental Mutagen Society (JEMS)-Mammalian Mutagenicity Study Group (MMS).
Mutation research (2005-05-19)
Hiroshi Suzuki, Naohiro Ikeda, Kazuo Kobayashi, Yukari Terashima, Yasushi Shimada, Takayoshi Suzuki, Toshiyuki Hagiwara, Shigeki Hatakeyama, Koko Nagaoka, Junichi Yoshida, Yukiko Saito, Jin Tanaka, Makoto Hayashi
RESUMEN
We conducted simultaneous liver and peripheral blood micronucleus assays in young rats with seven rodent hepatocarcinogens-4,4'-methylenedianiline (MDA), quinoline, o-toluidine, 4-chloro-o-phenylenediamine (CPDA), dimethylnitrosamine (DMN), p-dimethylaminoazobenzene (DAB), and di(2-ethylhexyl)phthalate (DEHP)-and two mutagenic chemicals-kojic acid and methylmethanesulfonate (MMS). Quinoline, DMN, and DAB were positive in the liver assay, while o-toluidine, kojic acid, DAB, and MMS were positive in the peripheral blood assay. o-Toluidine, kojic acid, and DAB are reportedly negative in mouse bone marrow micronucleus assays, indicating a species difference. Our results revealed a correlation between micronucleus induction in hepatocytes and hepatocarcinogenicity. This technique can be useful for the detection of micronucleus-inducing chemicals that require metabolic activation, and it enables simultaneous comparison of the micronucleus-inducing potential of chemicals in the liver and peripheral blood in the same individual.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Chloro-o-phenylenediamine, 97%