Merck
Pharmacological studies in animals of beta-hydroxyethyltheophylline, the major metabolite of doxofylline in humans.
Methods and findings in experimental and clinical pharmacology (1991-05-01)
J S Franzone, R Cirillo, M C Reboani
RESUMEN
beta-HET (beta-Hydroxyethyltheophylline), the major metabolite of the antibronchospastic, antiasthmatic drug doxofylline was studied in several in vitro and in vivo trials to characterize its pharmaco-toxicological profile. When compared to the parent compound, beta-HET was found to be significantly less active. It was also discovered to be a very weak inhibitor of phosphodiesterase activity. Its affinity for A1- and A2-adenosine receptors was even lower than that of doxofylline, which was quite low. The oral toxicity of beta-HET was about three times lower than that of doxofylline. The pharmacological activity of doxofylline is due to the drug in its original form and not to its major metabolite.