Merck
[Analysis of constituents of essential oil from the skin of water caltrop].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2006-05-26)
Rui Liang, Qi-Jun Peng
RESUMEN
To analyze the constituents of essential oil from the skin of water caltrop. Water steam distillation and GC-MS were used. 58 componds were separated respectively. 56 componds being identified which were 96. 5% of the totle essential oil. Diethyl phthalate, acetamide, N-acetyl-N, N'-1,2-ethanediylbis-, isopropyl palmitate, hexadecanoic acid, Z-11 and octadecanoic acid are the main component of essential oil from the skin of water caltrop.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Isopropyl palmitate, technical grade, 90%
Sigma-Aldrich
Isopropyl palmitate, ≥90%
Supelco
Isopropyl palmitate, Pharmaceutical Secondary Standard; Certified Reference Material