Merck
Susceptibilities of apple aphid and spirea aphid collected from apple in the Pacific Northwest to selected insecticides.
Journal of economic entomology (2006-08-30)
D Thomas Lowery, Michael J Smirle, Robert G Foottit, Elizabeth H Beers
RESUMEN
Laboratory bioassays using leaf disks of apple dipped in test solutions of insecticides demonstrated that the apple aphid, Aphis pomi De Geer, and the spirea aphid, Aphis spiraecola Patch, differed significantly in susceptibility to a number of insecticides registered for control of aphids on apple (Malus spp.). Compared with A. pomi, A. spiraecola was approximately four- and three-fold less susceptible to pirimicarb and lambda-cyhalothrin, respectively, whereas there was little difference in response to dimethoate. Pymetrozine is thought to act on aphids primarily as a feeding inhibitor. Exposure of aphids to this material generated data that fit the probit model for only half the tested clones. However, the LC50 value for one clone of A. spiraecola was nearly 1,000 times higher than the value for one clone of A. pomi. Although the results from these trials did not indicate that either species had developed significant levels of resistance to the test materials, differences in LC50 levels of > 10-fold suggest insecticide tolerances and the possibility of control failures in the future. The demonstrated differences in susceptibility to insecticides between these two morphologically similar species also should be considered during the evaluation of new products for use on apple.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Pymetrozin, PESTANAL®, analytical standard