Merck
  • Speciation of Zn-aminopolycarboxylic complexes by electrospray ionization mass spectrometry and ion chromatography with inductively coupled plasma mass spectrometry.
Speciation of Zn-aminopolycarboxylic complexes by electrospray ionization mass spectrometry and ion chromatography with inductively coupled plasma mass spectrometry.
Rapid communications in mass spectrometry : RCM (2009-01-08)
ZuLiang Chen, Gary Owen, Mallavarapu Megharaj, Ravendra Naidu
RESUMEN
The speciation of Zn-aminopolycarboxylic complexes was investigated using both electrospray ionization mass spectrometry (ESI-MS) and ion chromatography (IC) with inductively coupled plasma mass spectrometry (ICP-MS). The resulting ESI mass spectra indicated that [Zn(HEDTA)](1-), [Zn(NTA)](1-), [Zn(EDTA)](2-) and [Zn(DTPA)](3-) were all simultaneously detected in solution; [Zn(NTA)](1-) exhibited the weakest intensity of all these Zn-aminopolycarboxylic complexes. IC/ICP-MS was also successfully used to separate Zn complexes by anion-exchange chromatography using a mobile phase containing 30 mM (NH(4))(2)HPO(4) at pH 7.5 giving reasonable resolution within 15 min. A weak peak attributable to the poor stability [Zn(NTA)](1-) ion was also observed using IC/ICP-MS. With the exception of [Zn(NTA)](1-), detection limits ranging from 0.5 to 1.0 microg/L were obtained and the proposed method was used for the determination of Zn aminopolycarboxylic complexes in soil solution.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, ~98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, BioXtra, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad