Merck
  • Optimization of solid-phase extraction using developed modern sorbent for trace determination of ametryn in environmental matrices.
Optimization of solid-phase extraction using developed modern sorbent for trace determination of ametryn in environmental matrices.
Journal of hazardous materials (2009-07-04)
A R Koohpaei, S J Shahtaheri, M R Ganjali, A Rahimi Forushani, F Golbabaei
RESUMEN
In recent years, there has been a significant increase in molecularly imprinted solid-phase extraction (MISPE) technique for the purification and clean-up of environmental samples. In this study, solid-phase extraction using the imprinted polymer has been optimized with the experimental design approach for a triazine herbicide, named ametryn with regard to the critical factors such as sample pH, sample concentration, sample flow-rate, sample volume, elution solvent, washing solvent and sorbent mass. These factors were evaluated statistically and also validated with spiked drinking water samples and showed a good reproducibility over six consecutive days as well as six within-day experiments. Also, in order to the evaluate efficiency of the optimized MISPE protocols, enrichment capacity, reusability and cross-reactivity of cartridges have been studied. Finally, a selective MISPE was successfully demonstrated for ametryn with a recovery of above 90% for spiked drinking water samples. It was concluded that the central composite design could prove beneficial for aiding the MIP and MISPE development.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Ametryn, PESTANAL®, analytical standard