Merck
  • The influence of chronic eicosanoid biosynthesis inhibition on life history of the greater waxmoth, Galleria mellonella and its ectoparasitoid, Bracon hebetor.
The influence of chronic eicosanoid biosynthesis inhibition on life history of the greater waxmoth, Galleria mellonella and its ectoparasitoid, Bracon hebetor.
Journal of insect physiology (2011-02-10)
Ender Büyükgüzel, Hasan Tunaz, David Stanley, Kemal Büyükgüzel
RESUMEN
Eicosanoids are oxygenated metabolites of three C20 polyunsaturated fatty acids, mainly arachidonic acid (AA; 20:4n-6), but also 20:3n-6 and 20:5n-3. Aside from their importance in biomedicine, eicosanoids act in invertebrate biology. Prostaglandins (PGs) influence salt and water transport physiology in insect rectal epithelia and in Malpighian tubules. PGs also influence a few insect behaviors, including releasing oviposition behavior and behavioral fever. Eicosanoids act in ovarian development and in insect immunity. Because eicosanoids act in several areas of insect biology, we posed the hypothesis that chronic inhibition of eicosanoid biosynthesis, in the absence of microbial challenge, can influence insect life table parameters, including developmental time, survival, adult longevity and parasitoid fecundity. Here we report that inhibiting eicosanoid biosynthesis throughout the larval life exerted minor influences on some life table parameters of the greater wax moth, Galleria mellonella and its ectoparasitoid, Bracon hebetor, however, the inhibitors strongly reduced the production and hatchability of the parasitoids' eggs. The significance of the work relates to the potentials of understanding and targeting eicosanoid systems as a platform for developing new technologies of insect pest management. As seen here, the impact of targeting eicosanoid systems is seen in crucial moments of insect life histories, such as reproduction or immune challenge rather than in overall larval development.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
6,7-Dihydroxycoumarin, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
1-Phenyl-3-pyrazolidinone, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad