Merck
[Simultaneous determination of chlormequat chloride and mepiquat chloride in plants by mobile phase ion chromatography].
Se pu = Chinese journal of chromatography (2011-06-11)
Xu Zhou, Jingang Xu, Zhidong Chen, Yan Zhu
RESUMEN
Ion-pair reagent was used as mobile phase for the simultaneous separation of chlormequat chloride and mepiquat chloride in plants. The rapid separation was performed on a Dionex IonPac NG1 guard column and a Dionex IonPac NS1 analytical column using 1.0 mL/min of the eluent mixture of 1.00 mmol/L nonafluoropentanoic acid (as ion-pair reagent) and 7% acetonitrile, and the detection was achieved by a suppressed conductivity detector. The method provided good resolution of the analyte peaks without any interference. The detection limits of chlormequat chloride and mepiquat chloride were 0.154 6 mg/L and 0.171 4 mg/L, respectively. The linear calibration curves were obtained in the range of 1 - 100 mg/L. The mean recoveries in the ranges of 96.06% - 104.6% for chlormequat chloride and 98.53% - 103.7% for mepiquat chloride were obtained in real samples. The method requires only simple sample preparation and the technique is suitable for routine quality control analysis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(2-Chloroethyl)trimethylammonium chloride, 98%
Sigma-Aldrich
Chlorocholine chloride, BioReagent, suitable for plant cell culture
Supelco
Mepiquat chloride, PESTANAL®, analytical standard
Supelco
Chlormequat chloride, PESTANAL®, analytical standard