Merck
Influence of canthaxanthin on broiler breeder reproduction, chick quality, and performance.
Poultry science (2011-06-16)
W Zhang, K Y Zhang, X M Ding, S P Bai, J M Hernandez, B Yao, Q Zhu
RESUMEN
To investigate the effect of canthaxanthin supplied via a maternal route on the production of both breeder hens and chickens, 270 Chinese Three-Yellow breeder hens were randomly divided into 2 groups consisting of 135 birds each (5 replicates of 27) for study. The breeder hens were fed either a basal diet or the basal diet supplemented with 6 mg of canthaxanthin/kg for 24 wk. At the end of the 24-wk breeder experiment, all hatching eggs laid in 5 consecutive days of each group were collected and incubated. For each breeder group, 100 newly hatched chicks (5 replicates of 20) were reared under environmentally controlled conditions for 21 d. Canthaxanthin supplementation resulted in the following outcomes: an enhancement of the serum total antioxidant capacity (TAC) of breeder hens (P = 0.029), a significant increase in the yolk colorimetric score of Roche Yolk Color Fan (RYCF; P < 0.001), and a significant improvement of the antioxidant status of the egg yolk (P < 0.05). The chicks that hatched from eggs laid by breeder hens fed the canthaxanthin supplementation diet demonstrated a higher pigmentation colorimetric score of RYCF for their shank skin (P < 0.05), and the antioxidant capacity of the newly hatched chicks was significantly increased (P < 0.05). Both of these positive effects on shank skin pigmentation colorimetric score of RYCF and antioxidant capacity were observed for at least 7 d posthatching, and the chicks that hatched from canthaxanthin-enriched eggs showed a lower mortality (0 vs. 4%) during the first 21 d posthatching. These findings support the hypothesis that canthaxanthin supplementation of the maternal diet enhances the protective capacity of tissues against oxidative stress in vivo, which might be beneficial for poultry producers.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Canthaxanthin (trans), analytical standard