Merck
  • Lower rim 1,3-di{4-antipyrine}amide conjugate of calix[4]arene: synthesis, characterization, and selective recognition of Hg2+ and its sensitivity toward pyrimidine bases.
Lower rim 1,3-di{4-antipyrine}amide conjugate of calix[4]arene: synthesis, characterization, and selective recognition of Hg2+ and its sensitivity toward pyrimidine bases.
The Journal of organic chemistry (2012-01-24)
Jayaraman Dessingou, Khatija Tabbasum, Atanu Mitra, Vijaya Kumar Hinge, Chebrolu P Rao
RESUMEN
The structurally characterized lower rim 1,3-di{4-antipyrine}amide conjugate of calix[4]arene (L) exhibits high selectivity toward Hg(2+) among other biologically important metal ions, viz., Na(+), K(+), Ca(2+), Mg(2+), Mn(2+), Fe(2+), Co(2+), Ni(2+), Cu(2+), Zn(2+), Cd(2+), Hg(2+), Pb(2+), and Ag(+) as studied by fluorescence, absorption, and ESI MS. L acts as a sensor for Hg(2+) by switch-off fluorescence and exhibits a lowest detectable concentration of 1.87 ± 0.1 ppm. The complex formed between L and Hg(2+) is found to be 1:1 on the basis of absorption and fluorescence titrations and was confirmed by ESI MS. The coordination features of the mercury complex of L were derived on the basis of DFT computations and found that the Hg(2+) is bound through an N(2)O(2) extending from both the arms to result in a distorted octahedral geometry with two vacant sites. The nanostructural features such as shape and size obtained using AFM and TEM distinguishes L from its Hg(2+) complex and were different from those of the simple mercuric perchlorate. L is also suited to sense pyrimidine bases by fluorescence quenching with a minimum detection limit of 1.15 ± 0.1 ppm in the case of cytosine. The nature of interaction of pyrimidine bases with L has been further studied by DFT computational calculations and found to have interactions through a hydrogen bonding and NH-π interaction between the host and the guest.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4-Dimethylaminoantipyrine, reactive nitrogen species scavenger