Merck
Expedient enantioselective synthesis of cermizine D.
Organic letters (2012-03-01)
Nagarathanam Veerasamy, Erik C Carlson, Rich G Carter
RESUMEN
An efficient enantioselective synthesis of cermizine D has been developed that exploits the use of a common intermediate to access over 85% of the carbon backbone. Key steps include an organocatalyzed heteroatom Michael addition, a diastereoselective alkylation with α-iodomethyl phenyl sulfide, a conjugate addition to a vinyl sulfone species, and a sulfone coupling/desulfurization sequence to join the two major subunits.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Divinyl sulfone, contains hydroquinone as inhibitor, ≥96%
En este momento no podemos mostrarle ni los precios ni la disponibilidad