Merck
Chlorophyllin for the control of Ichthyophthirius multifiliis (Fouquet).
Parasitology research (2012-04-06)
S Wohllebe, P Richter, D-P Häder
RESUMEN
Tests were conducted to determine the efficiency of the photodynamic substance chlorophyllin to kill different life stages of the protozoan parasite Ichthyophthirius mulftifiliis (Fouquet) which causes the white spot disease in many freshwater fish species. This study has developed a new treatment for the control of ichthyophthiriosis, and demonstrated that non-toxic water-soluble chlorophyll (chlorophyllin) is a potential remedial agent at low concentrations for this serious parasitic disease.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Chlorophyllin sodium copper salt