Merck
A gold(I)-catalyzed route to α-sulfenylated carbonyl compounds from propargylic alcohols and aryl thiols.
Chemical communications (Cambridge, England) (2012-05-25)
Srijit Biswas, Joseph S M Samec
RESUMEN
A one-step atom efficient gold(I)-catalyzed route to α-sulfenylated ketones and aldehydes from propargylic alcohols and aryl thiols is described.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Propargyl alcohol, 99%