Merck
Biodegradability enhancement of wastewater containing cefalexin by means of the electro-Fenton oxidation process.
Journal of hazardous materials (2012-06-06)
Adriana Ledezma Estrada, Yu-You Li, Aimin Wang
RESUMEN
The comparative degradation behavior of cefalexin (CLX) using advanced oxidation processes (AOPs) with the aim of improving CLX biodegradability was studied. Among the AOPs used, RuO(2)/Ti anodic oxidation (AO), AO in the presence of electro-generated H(2)O(2) (AO-H(2)O(2)), and the electro-Fenton (EF) process, the EF process was the most effective. In the EF process an activated carbon fiber (ACF) was used as a cathode. Different input variables, including catalyst concentration, pH, and current density were evaluated to find the optimum conditions for the EF. The most suitable operational conditions were as follows: a current density of 6.66 mA/cm(2), a pH of 3, and a concentration of 1 mM of Fe(2+) as the catalyst. Different CLX concentrations were analyzed as well of different reaction times to assess the degree of mineralization. The change in biodegradability was evaluated by the BOD(5)/COD and the BOD(14)/COD ratios. The EF did not effectively remove the COD, but removed enough to achieve suitable biodegradability for a further biological process.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cephalexin hydrate, first-generation cephalosporin antibiotic
Supelco
Cefalexin, VETRANAL®, analytical standard
Cephalexin monohydrate, European Pharmacopoeia (EP) Reference Standard