Merck
[Tissue-specific expression of hormonal carcinogenesis target genes in rats treated with polycyclic aromatic hydrocarbons].
Biomeditsinskaia khimiia (2012-08-04)
M D Chanyshev, V O Pustyl'niak, L F Guliaeva
RESUMEN
We have investigated the effect of polycyclic aromatic hydrocarbons (PAHs) on estrogen-metabolizing genes CYP1A1, CYP1B1, CYP19 and ERalpha and cyclin D1 genes, which control of cell division in estrogen-depended tissues. Treatment of rats with benzo(a)pyren (BP) or 3-methylcholantrene (MC) significantly up-regulated CYP1A1, CYP1B1 gene expression in liver, uterus and ovary, whereas alfa-naphthoflavone (alpha-NF) did not have any effect. The high level of aromatase gene (CYP19) expression was detected in ovary only. Treatment of rats with BP or MC significantly down-regulated expression of this gene (15- and 5,5-fold, respectively), whereas alpha-NF did not have any effect. BP produced an increase in ERalpha and cyclin D1 gene expression in rat liver. This effect was not seen with MC and alpha-NF. ERalpha and cyclin D1 mRNA levels were unchanged in uterus of rats after PAHs treatment. On the other hand, BP treatment caused an increase of the ERalpha and cyclin D1 mRNA levels (3,5- and 2,5-fold, respectively) in ovary, whereas MC and alpha-NF did not have any effects. Thus, our results give evidence for tissue-specific effects of PAHs on expression of genes, which participate in hormonal carcinogenesis. Moreover, the fact that BP and MC treatment affects the expression of estrogen-metabolizing genes and genes, which control of cell division, supports the view that PAHs may be one of the causes of endocrine disorder and consequent hormonal carcinogenesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3-Methylcholanthrene (20), 98%
Supelco
3-Methylcholanthrene (20), analytical standard
Sigma-Aldrich
α-Naphthoflavone, ≥98%
Supelco
3-Methylcholanthrene solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard