Merck
Asymmetric iodolactonization utilizing chiral squaramides.
Organic letters (2012-11-15)
Jørn E Tungen, Jens M J Nolsøe, Trond V Hansen
RESUMEN
Asymmetric iodolactonization of γ- and δ-unsaturated carboxylic acids has been explored in the presence of six different chiral organocatalysts 5-8. The catalyst 6b was found to facilitate the cyclization of 5-arylhex-5-enoic acids 1 to the corresponding iodolactones 2 with up to 96% ee. By this protocol, unsaturated carboxylic acids are converted enantioselectively to synthetically useful δ-lactones in high yields using commercially available NIS. Apparently, both hydrogen bonding and aryl/aryl interactions are important for efficient stereodifferentiation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)