Merck
The occurrence of semicarbazide in the meat and shell of Bangladeshi fresh-water shrimp.
Food chemistry (2012-12-01)
Robert McCracken, Bob Hanna, David Ennis, Lynne Cantley, Dermot Faulkner, D Glenn Kennedy
RESUMEN
There is evidence that semicarbazide (SEM), a marker for the banned nitrofuran nitrofurazone, can arise from other, unrelated sources. Recently, Belgium rejected 54 consignments of Bangladeshi freshwater shrimp (Macrobrachium rosenbergii), following a laboratory decision to test meat and exoskeleton combined. To study the possible natural occurrence of SEM in wild shrimp, samples were collected and analysed from 29 sites across Bangladesh. SEM (<1.0 μg/kg) was detected in ∼65% of meat samples. However, SEM concentrations were approximately 100 times higher in the exoskeleton, and were unrelated to sampling location, strongly suggesting natural occurrence. In meat, most SEM was surface-associated. When the shrimp was shelled, some of the epidermal layer (which synthesises new exoskeleton) remained with the shell and some remained with the meat--leading to differing levels of natural SEM on the shrimp surface. This has implications for the use of SEM and the analytical strategy used to control nitrofuran use.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Semicarbazide hydrochloride, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Semicarbazide, 6 wt. % (on silica gel)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Semicarbazide hydrochloride, VETRANAL®, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad